Acros Organics Chloroform

Quality and consistency your lab can count on
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$47.75 - $183.00USD / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Acros Organics AC158210025 Chloroform, stabilized 99+% (2.5l) EW-88164-21 Mfr # AC158210025
COMPARE
Stabilized 2500 Chloroform, stabilized 99 % (2.5 L)
$150.00USD / Each
Acros Organics AC167735000 Chloroform, stabilized, spectrophotometricgrade 99+% (500ml) EW-88177-90 Mfr # AC167735000
COMPARE
Spectrophotometric 500 Chloroform, stabilized, spectrophotometricgrade 99 % (500 mL)
$71.50USD / Each
Acros Organics AC232090025 Chloroform, stabilized p.a. (2.5l) CAS 67-66-3 EW-88247-12 Mfr # AC232090025
COMPARE
Pro Analysi 2500 Chloroform, stabilized p.a. (2.5 L)
$183.00USD / Each
Acros Organics AC268320025 Chloroform, stabilized, for HPLC (2.5l) EW-88258-94 Mfr # AC268320025
COMPARE
HPLC 2500 Chloroform, stabilized, for HPLC (2.5L)
$175.00USD / Each
Acros Organics AC364320010 Chloroform, extra dry, with molecular sieves (1l) EW-88337-64 Mfr # AC364320010
COMPARE
1000 Chloroform, extra dry, with molecular sieves (1 L)
$92.50USD / Each
Acros Organics AC404635000 Chloroform, reagent ACS, spectro grade (500 mL) EW-88341-97 Mfr # AC404635000
COMPARE
ACS Reagent or Spectrophotometric 500 Chloroform, Reagent ACS, spectro grade (500 mL)
$73.50USD / Each
Acros Organics AC610281000 Chloroform, stabilized with amylene, anhydrous, 99.9%, AcroSeal (100 mL) EW-88364-45 Mfr # AC610281000
COMPARE
Anhydrous 100 Chloroform, stabilized with amylene, anhydrous, 99.9%, AcroSeal (100 mL)
$47.75USD / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat