Acros Organics Hexanes

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$104.75 - $348.75USD / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
Acros Organics n-Hexane for Analysis, 99+%, 1 L EW-88168-45 Mfr # AC160780010
COMPARE
n-Hexane 1000 n-Hexane for Analysis, 99+%, 1 L
$244.00USD / Each
Acros Organics Hexanes, Extra Pure, Mixture of Isomers, 98+%, 1 L EW-88189-51 Mfr # AC176660010
COMPARE
Hexanes 1000 Hexanes, Extra Pure, Mixture of Isomers, 98+%, 1 L
$104.75USD / Each
Acros Organics Hexanes, Extra Pure, Mixture of Isomers, 98+%, 2.5 L EW-88189-52 Mfr # AC176660025
COMPARE
Hexanes 2500 Hexanes, Extra Pure, Mixture of Isomers, 98+%, 2.5 L
$181.25USD / Each
Acros Organics n-Hexane, Extra Pure, 95+%, 1 L EW-88211-01 Mfr # AC197360010
COMPARE
n-Hexane 1000 n-Hexane, Extra Pure, 95+%, 1 L
$146.75USD / Each
Acros Organics n-Hexane, Extra Dry over Molecular Sieve, 97%, 1 L EW-88337-74 Mfr # AC364370010
COMPARE
n-Hexane 1000 n-Hexane, Extra Dry over Molecular Sieve, 97%, 1 L
$210.50USD / Each
Acros Organics Hexanes, ACS Reagent, Mixture of Isomers, 99%, 500 mL EW-88348-58 Mfr # AC411555000
COMPARE
Hexanes 500 Hexanes, ACS Reagent, Mixture of Isomers, 99%, 500 mL
$118.00USD / Each
Acros Organics Hexanes, HPLC, 98.5%, 4 L EW-88364-27 Mfr # AC610070040
COMPARE
Hexanes 4000 Hexanes, HPLC, 98.5%, 4 L
$348.75USD / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat