This product is now discontinued, View Related Products

Acros Organics n-Hexane

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Acros Organics AC160780025 n-Hexane p.a. (2.5l) 8816846 EW-88168-46 Mfr # AC160780025
COMPARE
110-54-3 2500 n-Hexane p.a. (2.5l) 8816846
Call For Price
Acros Organics AC160780040 n-Hexane p.a. (4l) 8816847 EW-88168-47 Mfr # AC160780040
COMPARE
110-54-3 4000 n-Hexane p.a. (4l) 8816847
Call For Price
Acros Organics AC197360250 n-Hexane 95% (25l) 8821103 EW-88211-03 Mfr # AC197360250
COMPARE
110-54-3 25000 n-Hexane 95% (25l) 8821103
Call For Price
Acros Organics AC326660010 n-Hexane, ECD tested for pesticide analysis, residue<0.0003% (1l) EW-88294-47 Mfr # AC326660010
COMPARE
110-54-3 1000 n-Hexane, ECD tested for pesticide analysis, residue<0.0003% (1l)
Call For Price
Acros Organics AC326660025 n-Hexane, ECD tested for pesticide analysis, residue<0.0003% (2.5l) EW-88294-48 Mfr # AC326660025
COMPARE
110-54-3 2500 n-Hexane, ECD tested for pesticide analysis, residue<0.0003% (2.5l)
Call For Price
Acros Organics AC326710010 n-Hexane, ECD tested halocarbons free grade (1l) EW-88294-57 Mfr # AC326710010
COMPARE
110-54-3 1000 n-Hexane, ECD tested halocarbons free grade (1l)
Call For Price
Acros Organics AC326710025 n-Hexane, ECD tested halocarbons free grade (2.5l) EW-88294-58 Mfr # AC326710025
COMPARE
110-54-3 2500 n-Hexane, ECD tested halocarbons free grade (2.5l)
Call For Price
Acros Organics AC326780010 n-Hexane, tested for trace analysis of polyaromatic hydrocarbons (1l) EW-88294-71 Mfr # AC326780010
COMPARE
110-54-3 1000 n-Hexane, tested for trace analysis of polyaromatic hydrocarbons (1l)
Call For Price
Acros Organics AC326780025 n-Hexane, tested for trace analysis of polyaromatic hydrocarbons (2.5l) EW-88294-72 Mfr # AC326780025
COMPARE
110-54-3 2500 n-Hexane, tested for trace analysis of polyaromatic hydrocarbons (2.5l)
Call For Price
Acros Organics AC326921000 n-Hexane, extra dry, water <20 ppm (100ml) EW-88295-01 Mfr # AC326921000
COMPARE
110-54-3 100 n-Hexane, extra dry, water <20 ppm (100ml)
Call For Price
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat