Acros Organics Sodium Bromide

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$93.20 - $283.75USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Weight (g) Description
Availability Pricing
Fisher Chemical S255500 Sodium Bromide, Granular, Certified ACS; 500 g EW-88023-24 Mfr # S255500
COMPARE
7647-15-6 500 Sodium Bromide (Granular/Certified ACS) (500g)
$213.25USD / Each
Acros Organics Sodium Bromide, Anhydrous, 99+%, Crystalline, Extra Pure Grade; 1 kg EW-88219-31 Mfr # AC205130010
COMPARE
7647-15-6 1000 Sodium bromide, anhydrous 99+% (1kg) CAS 7647-15-6
$283.75USD / Each
Acros Organics AC246905000 Sodium Bromide, p.a., 99.5%, Crystalline, Analysis Grade; 500 g EW-88250-90 Mfr # AC246905000
COMPARE
7647-15-6 500 Sodium bromide p.a. (500g) CAS 7647-15-6
$134.00USD / Each
COMPARE
7647-15-6, 500 Sodium Bromide, ACS Grade, 500 g
$93.20USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat