Acros Organics Tetrhydrofuran

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$122.00 - $450.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Acros Organics Tetrahydrofuran, 99+%, Stabilized; 1 L EW-88189-46 Mfr # AC176630010
COMPARE
Stabilized 1000 Tetrahydrofuran, stabilized 99+% (1 L)
$135.00USD / Each
Acros Organics Tetrahydrofuran, 99+%, Stabilized; 2.5 L EW-88189-47 Mfr # AC176630025
COMPARE
Stabilized 2500 Tetrahydrofuran, stabilized 99+% (2.5 L)
$233.75USD / Each
Acros Organics Tetrahydrofuran, HPLC Grade; 2.5 L EW-88258-85 Mfr # AC268290025
COMPARE
HPLC 2500 Tetrahydrofuran, for HPLC (2.5 L)
$450.00USD / Each
Acros Organics Tetrahydrofuran, Water < 50 ppm, Extra Dry over Molecular Sieve Grade; 100 mL EW-88318-21 Mfr # AC348451000
COMPARE
Extra Dry 100 Tetrahydrofuran, extra dry, with molecularsieve, water < 50 ppm (100 mL)
$122.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat