Acros Organics Titanium

Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$53.30 - $269.25USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Class Form Description
Availability Pricing
Acros Organics Titanium(IV) isopropoxide 98+% (250 mL) CAS 546-68-9 EW-88207-06 Mfr # AC194702500
COMPARE
General Solutions Liquid Titanium(IV) isopropoxide 98+% (250 mL) CAS 546-68-9
$126.75USD / Each
Acros Organics Titanium(IV) chloride 99.9% (100 mL) CAS 7550-45-0 EW-88210-79 Mfr # AC197231000
COMPARE
Reagent Salts Liquid Titanium(IV) chloride 99.9% (100 mL) CAS 7550-45-0
$269.25USD / Each
Acros Organics Titanium(IV) oxide, anatase powder 99% (100g) EW-88228-13 Mfr # AC213581000
COMPARE
Solids Powder Titanium(IV) oxide, anatase powder 99% (100g)
$53.30USD / Each
Acros Organics Titanium(IV) n-butoxide 99% (500g) EW-88239-14 Mfr # AC223195000
COMPARE
General Solution Liquid Titanium(IV) n-butoxide 99% (500g)
$190.50USD / Each
Acros Organics Titanium(IV) oxide 98.0 - 100.5% TiO2 (1kg) EW-88264-32 Mfr # AC277370010
COMPARE
Solids Solid Titanium(IV) oxide 98.0 - 100.5% TiO2 (1kg)
$112.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat