Acrylic Balance Enclosure
  • Protects you from dust, particulates, and organic vapor
Variable-speed blower allows continuous adjustment of airflow and face velocity. Front hinged panel is constructed of clear polycarbonate and allows esy access to work area. Order the optional fluorescent light kit if additional lighting is needed. Optional audible alarms notify you in the event of an airflow failure or a blocked filter.
Shipping weight: 80 lb (36.3 kg)
Supplied HEPA filter removes particles down to 0.3 microns at 99.99% efficiency. When working with large particulate samples, purchase a prefilter to prevent premature clogging of the HEPA filter. Optional carbon filters are available for the adsorption of specific vapors; order separately. Units include a 6-ft power cord with three-prong plug.
$3,065.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Exterior Width (in) Power (VAC)
Availability Pricing
Airfiltronix BE-1 Balance Enclosure, acrylic, 115 VAC EW-44011-00 Mfr # BE-1 115V
COMPARE
34.5 30 115
$3,065.00USD / Each