Plas-Labs Acrylic Desiccators with Gas Ports
Gas ports allow you to create an oxygen-free or inert gas atmosphere
  • Humidity levels are monitored with easy-to-read built-in hygrometer
  • Slip-away hinges allow complete removal of door when needed
  • Amber-colored desiccators protect from UV light
These heavy-duty desiccators are made of 1⁄4"-thick acrylic for durability. Resilient, closed-cell PVC gaskets prevent system leaks. Temperature range is 40 to 120°F (4 to 49°C). All models feature two gas ports—1⁄4" hose barb inlet; 1⁄4" hose barb outlet. Choose clear units for maximum visibility of contents or amber units for protecting lightsensitive compounds from ultraviolet light. All models are easy to access and clean; slip-away hinges allow complete removal of door when needed. Adjust shelf positions to suit your needs. Models 08900-54 thru -64 have a nonremovable divider separating the chamber into two separate cubicles.
$737.00 - $2,063.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Exterior Depth (in) Exterior Height (in)
Availability Pricing
Plas-Labs 860-CG Acrylic Desiccator with Gas Port, Clear; 12"W x 12"H x 12"D EW-08900-50 Mfr # 860-CG/NS
COMPARE
Clear 12 12
$737.00USD / Each
Plas-Labs 860-CGA Acrylic Desiccator with Gas Port, Amber; 12"W x 12"H x 12"D EW-08900-52 Mfr # 860-CGA/NS
COMPARE
Amber 12 12
$763.50USD / Each
Plas-Labs 861-CG Acrylic Desiccator with Gas Port, Clear; 12"W x 24"H x 12"D EW-08900-54 Mfr # 861-CG/NS
COMPARE
Clear 12 24
$1,211.00USD / Each
Plas-Labs 861-CGA Acrylic Desiccator with Gas Port, Amber; 12"W x 24"H x 12"D EW-08900-56 Mfr # 861-CGA/NS
COMPARE
Amber 12 24
$1,313.00USD / Each
Plas-Labs 862-CG Acrylic Desiccator with Gas Port, Clear; 18"W x 18"H x 18"D EW-08900-58 Mfr # 862-CG/NS
COMPARE
Clear 18 18
$1,590.00USD / Each
Plas-Labs 862-CGA Acrylic Desiccator with Gas Port, Amber; 18"W x 18"H x 18"D EW-08900-60 Mfr # 862-CGA/NS
COMPARE
Amber 18 18
$1,747.00USD / Each
Plas-Labs 863-CG Acrylic Desiccator with Gas Port, Clear; 24"W x 24"H x 18"D EW-08900-62 Mfr # 863-CG/NS
COMPARE
Clear 18 24
$1,832.00USD / Each
Plas-Labs 863-CGA Acrylic Desiccator with Gas Port, Amber; 24"W x 24"H x 18"D EW-08900-64 Mfr # 863-CGA/NS
COMPARE
Amber 18 24
$2,063.00USD / Each
Showing 1- 8 of 8