Adam Equipment Output / Relay Kits
Quickly and easily program up to four switches
  • Program four switches to operate with the checkweighing functions
  • Relays can be set up to drive 115/230VAC 1A or 24VDC 1A output
The relay box is an add-on for the ABK, AFK, AGB, AGF, and Warrior series industrial scales, as well as the AE 402 and AE 403 scale indicators. When installed, it allows the checkweighing limits to trigger a voltage relay. Once the trigger is set, the relay box will open or close a switch. This allows the scales to drive other items such as a motor for filling, or signal lights. The relay box includes input for power to help drive the relays.
A light tower kit with relay box is available to notify you at a glance if your weight sample is over, under, or within the preset limits.
ITEM(S) ON SALE
$166.25USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accessory Type
Availability Pricing
Adam Equipment Relay Output Box for Scale Indicator EW-11710-35 Mfr # 1100011297
COMPARE
Relay output box
$166.25USD / Each
$190.00USD / Each