Additel 680W Wireless Digital Pressure Gauges with Data Logging
No need to remove gauge from service to download data
  • 2.4 GHz ISM bands communicate with your computer wirelessly from up to 66 ft (20 m) away
  • IP67-rated construction withstands a 3-ft (1-m) submersion in water or a drop of 3 ft (1 m)
  • Large, backlit 5-digit display shows pressure, % of full-scale graph, and temperature
  • Icon-based programming menu is easy to navigate
Additel 680W digital pressure gauges use an advanced technology microprocessor and are aged, tested, and screened before assembly. With data logging and wireless capabilities, these gauges allow you to monitor and log pressure for real-time monitoring and logging to a PC wirelessly. Gauge can store up to 140,000 readings of pressure, temperature, and time.
Supplied software allows reviewing of data after logging period is ended. Read as many gauges as you'd like on one PC by setting the same channel (1 to 15) on the gauge as the PCs' software, or read various gauges on different PCs by synching the channels to match PC and gauge. Optional software (order separately) shows current data being logged in real time on your PC and allows graphical analysis.
$874.00 - $1,533.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Min Pressure (PSI)
Availability Pricing
Additel Digital Pressure Gauge with Wireless RF Transmitter, -15 to 0 psi EW-68410-20 Mfr # ADT680W-25-V15-PSI-N
COMPARE
±0.25% full-scale 0 -15
$874.00USD / Each
Additel Digital Pressure Gauge with Wireless RF Transmitter, 0 to 15 psi EW-68410-26 Mfr # ADT680W-25-GP15-PSI-N
COMPARE
±0.25% full-scale 15 0
$874.00USD / Each
Additel Digital Pressure Gauge with Wireless RF Transmitter, 0 to 30 psi EW-68410-28 Mfr # ADT680A-20-GP30-PSI-N
COMPARE
±0.25% full-scale 30 0
$874.00USD / Each
Additel Digital Pressure Gauge with Wireless RF Transmitter, 0 to 100 psi EW-68410-30 Mfr # ADT680W-25-GP100-PSI-N
COMPARE
±0.25% full-scale 100 0
$874.00USD / Each
Additel Digital Pressure Gauge with Wireless RF Transmitter, 0 to 300 psi EW-68410-32 Mfr # ADT680W-25-GP300-PSI-N
COMPARE
±0.25% full-scale 300 0
$874.00USD / Each
Additel Digital Pressure Gauge with Wireless RF Transmitter, 0 to 1000 psi EW-68410-36 Mfr # ADT680A-20-GP1K-PSI-N
COMPARE
±0.25% full-scale 1000 0
$874.00USD / Each
Additel Digital Pressure Gauge with Wireless RF Transmitter, 0 to 10,000 psi EW-68410-42 Mfr # ADT680W-25-GP10K-PSI-N
COMPARE
±0.25% full-scale 10000 0
$948.00USD / Each
Additel Digital Pressure Gauge with Wireless RF Transmitter, 0 to 15,000 psi EW-68410-44 Mfr # ADT680W-25-GP15K-PSI-N
COMPARE
±0.25% full-scale 15000 0
$1,113.00USD / Each
Additel Digital Pressure Gauge with Wireless RF Transmitter, 0 to 25,000 psi EW-68410-46 Mfr # ADT680W-25-GP25K-PSI-AM
COMPARE
±0.25% full-scale 25000 0
$1,324.00USD / Each
Additel Digital Pressure Gauge with Wireless RF Transmitter, 0 to 30,000 psi EW-68410-48 Mfr # ADT680W-25-GP30K-PSI-AM
COMPARE
±0.25% full-scale 30000 0
$1,533.00USD / Each
Showing 1- 10 of 10