Additel Loop Calibrators
Easy, ergonomic one-handed operation
  • Simulate, source and measure a variety of signals
  • HART® communications with internal resistor standard on both models
  • Selectable ramp and step functions allow calibration of 4 to 20 mA devices
Large, high-contrast VA LCD displays both measurement and percent of span. Functions include DC measurement (V, mA), switch test (input resistance >500 M ohm) with low and high level triggers, and source or simulate (mA). User-accessible diagnostics and calibration menus let you maintain your device.
$925.00 - $1,218.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Additel 209 Current Loop Calibrator, HART Communications, Standard Accuracy EW-16102-22 Mfr # ADT209
COMPARE
Current Loop Calibrator, HART Communications, Standard Accuracy
$925.00USD / Each
Additel 210 Current Loop Calibrator, HART Communications, High Accuracy EW-16102-24 Mfr # ADT210
COMPARE
Current Loop Calibrator, HART Communications, High Accuracy
$1,218.00USD / Each