Additel Pneumatic and Hydraulic Pressure Manifolds
Expand productivity for high throughput pressure calibration applications
  • Hand-tightening quick connectors eliminate need for wrenches and PTFE tape
  • Built-in filter on pneumatic models prevents migration of contaminants
  • Calibrate a variety of instruments using models with adjustable manifold arms
  • Choose between pneumatic and hydraulic models with pressure ranges up to 15,000 psi
Quickly change out devices under test, calibration pumps, dead weight testers, and pressure controllers with the hand-tightened quick connectors. Use the adjustable-arm models to alternate between a space-saving compact footprint for small gauges and transducers, and a more spread out orientation to accommodate larger gauges.
$1,558.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Min Pressure
Availability Pricing
Additel ADT121-N Pneumatic Pressure Manifold, -15 to 3000 psi, 1/4" NPT (F) EW-16107-93 Mfr # ADT121-N
COMPARE
3000 psi -15 psi
$1,558.00USD / Each
Additel ADT123-N Pneumatic Pressure Manifold, -15 to 15000 psi, 1/4" NPT (F) EW-16107-94 Mfr # ADT123-N
COMPARE
15000 psi -15 psi
$1,558.00USD / Each