Advanced Green 3

Advanced Green 3 are low-bleed septa that are specifically created to combine a long injection life with low injection port adhesion to meet even your most challenging analysis. Maximum temperature 305°C. Pre-piercing eliminates injection coring on select CenterGuide septa.
$92.50 - $107.00USD / Pkg of 50

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Advanced Green 3 246124 Septa, 3/8" (9.5 mm); 50/Pack EW-01994-50 Mfr # 246124
COMPARE
Septa, 3/8" (9.5 mm); 50/Pack
$102.00USD / Pkg of 50
Advanced Green 3 246225 Septa, 7/16" (11 mm); 50/Pack EW-01994-52 Mfr # 246225
COMPARE
Septa, 7/16" (11 mm); 50/Pack
$106.00USD / Pkg of 50
Advanced Green 3 246624 Septa, 11/16" (17 mm); 50/Pack EW-01994-56 Mfr # 246624
COMPARE
Septa, 11/16" (17 mm); 50/Pack
$107.00USD / Pkg of 50
Advanced Green 3 246713 Septa, CenterGuide, 9 mm; 50/Pack EW-01999-20 Mfr # 246713
COMPARE
Septa, CenterGuide, 9 mm; 50/Pack
$106.00USD / Pkg of 50
Advanced Green 3 Septa, CenterGuide, 11.5 mm; 50/Pack EW-01999-22 Mfr # 246725
COMPARE
Septa, CenterGuide, 11.5 mm; 50/Pack
$92.50USD / Pkg of 50
Advanced Green 3 246424 Septa, Shimadzu; 50/Pack EW-01999-24 Mfr # 246424
COMPARE
Septa, Shimadzu; 50/Pack
$93.50USD / Pkg of 50
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat