This product is now discontinued, View Related Products
AEMC AC-DC Low Current Probe
FEATURES
  • Measures extremely low level DC from 100µA
  • Outputs signal proportional to total current (DC + AC)
  • Low noise
  • Ultra compact size and non contact clamp on convenience
  • Simple plug in operation
  • Designed for use with digital multimeters and oscilloscopes
  • Accurate display of waveforms
  • No range or mode (AC/DC) switching required
  • Extended battery life
  • Red LED indicates momentary or continuous overload
  • Green LED indicates power and battery condition
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
AEMC K100 AC/DC Low Current MicroProbe, 0 to +/-4.5 A EW-20002-07 Mfr # 1200.67
COMPARE
Call For Price
AEMC K110 DC/AC Microprobe Low Current Probe 10mV/Ma Output EW-20002-29 Mfr # 2111.73