AEMC AmpFlex Flexible Current Probes
These AmpFlex ® flexible AC current probes are composed of a flexible sensor and an electronic module. The flexible sensor permits measurements on conductors where standard clamp-on probes cannot be used. In particular, it can be installed in tight spaces, around cable bundles, around wide or large bus bars, or even wrapped around irregular shapes. The Shape Memory feature enables the user to pre-shape the sensor before inserting in between or around the conductor, enhances user safety, and alleviates the drooping effect associated with other flexible sensors.
FEATURES
  • Measure from 30Arms to 30,000 Arms
  • Lengths from 24" to 60" standard
  • Accuracy 1% of read
  • 1 PRODUCT OPTIONS
    Compareitem specifications
    DC Max (Volts) DC Voltage Accuracy True Rms
    Availability Pricing
    AEMC 1000-24-2-1 AmpFlex flexible current probe 100/1000 A, 24", 10 mV/1 mV/A EW-20002-33 Mfr # 2112.98
    COMPARE
    $437.00USD / Each