AEMC Cable Testers
  • Fault Mapper
    Cable Length Meter and Fault Locator Alphanumeric TDR - AE212780
  • Handheld cable length meter and fault locator
  • Detects opens and shorts and
  • the distance to them from one end of the cable
  • Measure cable length up to 6000 ft or 2000m (selectable)
  • Automatic cable impedance compensation
  • Built-in library of most common cables and their Velocity of Propagation (Vp)
  • Manual selection of Vp for all cables not found in library
  • User programmable cable library with 15 positions
  • Vp setting displayed along with length and cable type (if in library)
  • Built-in tone generator for tracing and locating cables
  • Large high-visibility blue electroluminescent backlit display
Wire Mapper Pro LAN Cable Tester, Fault Mapper Cable Length Meter and Fault Locator Alphanumeric TDR, and Fault Mapper Pro Telephone / Coaxial / Parallel Cable Tester
$599.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Input Current (mA) Max Input Voltage (VDC) Number of Ports
Availability Pricing
AEMC CA7024 Fault Mapper Alpha Numeric TDR EW-17782-01 Mfr # 2127.8
COMPARE
$599.00USD / Each
AEMC CA7028 Wire Mapper Pro Cable Tester EW-17782-03 Mfr # 2127.82
COMPARE
$599.00USD / Each