AEMC Phase and Rotation Meters
Determine correct wiring for proper rotation on motors, pumps, and more
  • Verify proper phase wiring
  • Detect open or de-energized phases
These testers are ideal for identifying the phase order on an electrical installation and the rotational direction of industrial equipment such as motors, pumps and valves connected to the installation. The color-coding of the terminals and cables on these testers makes set up and use simple. Model 6608 can be used to quickly determine the phase order and detect any phase loss.
Model 6609 includes additional functions specially designed for industrial equipment (motors, pumps, etc) that include indication of the rotating field direction without connecting any test leads, and determination of the connection and direction of each phase.
$94.00 - $124.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
AEMC 2121.10 40 to 850 Vac Phase Rotation Meter EW-20035-80 Mfr # 6608
COMPARE
850
$94.00USD / Each
AEMC 2121.11 Phase and Motor Rotation Meter EW-20035-81 Mfr # 6609
COMPARE
600
$124.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat