Air Cleaning Systems
Two self-supporting extractor arms are ideal for two-station setups
These air cleaning systems capture and remove smoke, dust, mist, and fumes before they enter the work area, then recirculate clean air back into the workplace. The systems are self-contained, with two 3"" dia self-supporting extractor arms made of heat-, chemical-, and abrasion-resistant polypropylene; end of arms flare up to 4 1/2"" dia. Ideal for two-station setups, or use the included inlet cap for single-arm use. Choose from bench top or portable system, then complete your system with filters sold separately.
Recommended for:
-Lab fumes and dust
-Clean room applications
-Soldering
-Small grinding applications
-Light welding
-Cleaning with solvents
-Art restoration & conservation
$1,975.00 - $2,753.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Exterior Width (in) Description
Availability Pricing
Air Impurities Removal Systems Air Cleaning System, benchtop, dual arm, 115 VAC EW-33703-25 Mfr # S-981-2B
COMPARE
10 12 Air Cleaning System, benchtop, dual arm, 115 VAC
$1,975.00USD / Each
Air Impurities Removal Systems Air Cleaning System, portable, dual arm, 115 VAC EW-33703-30 Mfr # SP-981-2B
COMPARE
36.8125 20.5 Air Cleaning System, portable, dual arm, 115 VAC
$2,753.00USD / Each