Air/N2 Gas Mass Flow Controllers with Metal bodies
Precisely control flow rates with built-in valve
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These controllers feature an advanced straight-tube sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity.
The flow rate set point can be established by either a local potentiometer or by a remote 4 to 20 mA or 0 to 5 VDC signal. Output data is sent via a 0 to 5 VDC or 4 to 20 mA signal; an analog-to-RS converter is also available for data collection and analysis on your PC.
All controllers include a detachable 3-1/2-digit LCD that can be tilted up to 90° for easy reading. The display is remote mountable to three feet using extension cable 32662-70. The totalizer option 32650-22 will display total accumulated flow.
The controller is protected from polarity reversal or short circuit by a built-in resettable fuse. Aluminum models have wetted materials of anodized aluminum, brass, and Viton; stainless steel (SS) models have wetted materials of SS and Viton.
Accuracy:
These precalibrated flow controllers operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77°F (15 to 25°C) while maintaining the stated percent full-scale accuracy and linearity. When operating beyond 5 to 60 psi, add ±0.01%/psi full-scale; if operating beyond 59 to 77°F (15 to 25°C), add ±0.15%/°C full-scale.
$941.00 - $2,428.00USD / Each
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Process Connection Wetted Materials
Availability Pricing
Aalborg Compact Gas Mass Flow Controller, 0-10 sccm, N2/Air, Aluminum Body
Buy More Save More
EW-32660-00 Mfr # GFCS-010002
COMPARE
0.01 1/4" Compression Anodized Aluminum, Brass, Viton
$941.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-20 sccm, N2/Air, Aluminum Body
Buy More Save More
EW-32660-02 Mfr # GFCS-010003
COMPARE
0.02 1/4" Compression Anodized Aluminum, Brass, Viton
$941.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-50 sccm, N2/Air, Aluminum Body
Buy More Save More
EW-32660-04 Mfr # GFCS-010004
COMPARE
0.05 1/4" Compression Anodized Aluminum, Brass, Viton
$941.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-100 sccm, N2/Air, Aluminum Body
Buy More Save More
EW-32660-06 Mfr # GFCS-010173
COMPARE
0.1 1/4" Compression Anodized Aluminum, Brass, Viton
$941.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-500 sccm, N2/Air, Aluminum Body
Buy More Save More
EW-32660-10 Mfr # GFCS-010007
COMPARE
0.5 1/4" Compression Anodized Aluminum, Brass, Viton
$941.00USD / Each
Aalborg GFCS Compact Gas Mass Flow Controller, 0-1 LPM, N2/Air, Aluminum Body
Buy More Save More
EW-32660-12 Mfr # GFCS-010008
COMPARE
1 1/4" Compression Anodized Aluminum, Brass, Viton
$941.00USD / Each
Aalborg GFCS Compact Gas Mass Flow Controller, 0-5 LPM, N2/Air, Aluminum Body
Buy More Save More
EW-32660-16 Mfr # GFCS-010010
COMPARE
5 1/4" Compression Anodized Aluminum, Brass, Viton
$941.00USD / Each
Aalborg GFCS Compact Gas Mass Flow Controller, 0-50 LPM, N2/Air, Aluminum Body
Buy More Save More
EW-32660-22 Mfr # GFCS-010013
COMPARE
50 1/4" Compression Anodized Aluminum, Brass, Viton
$1,117.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-100 LPM, N2/Air, Aluminum Body
Buy More Save More
EW-32660-24 Mfr # GFCS-010014
COMPARE
100 3/8" Compression Anodized Aluminum, Brass, Viton
$1,290.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-50 sccm, N2/Air, Stainless Steel Body
Buy More Save More
EW-32661-04 Mfr # GFCS-010056
COMPARE
0.05 1/4" Compression Stainless Steel, Viton
$1,087.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-100 sccm, N2/Air, Stainless Steel Body
Buy More Save More
EW-32661-06 Mfr # GFCS-010057
COMPARE
0.1 1/4" Compression Stainless Steel, Viton
$1,087.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-500 sccm, N2/Air, Stainless Steel Body
Buy More Save More
EW-32661-10 Mfr # GFCS-010059
COMPARE
0.5 1/4" Compression Stainless Steel, Viton
$1,087.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-1 LPM, N2/Air, Stainless Steel Body
Buy More Save More
EW-32661-12 Mfr # GFCS-010060
COMPARE
1 1/4" Compression Stainless Steel, Viton
$1,087.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-2 LPM, N2/Air, Stainless Steel Body
Buy More Save More
EW-32661-14 Mfr # GFCS-010061
COMPARE
2 1/4" Compression Stainless Steel, Viton
$1,087.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-5 LPM, N2/Air, Stainless Steel Body
Buy More Save More
EW-32661-16 Mfr # GFCS-010062
COMPARE
5 1/4" Compression Stainless Steel, Viton
$1,087.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-15 LPM, N2/Air, Stainless Steel Body
Buy More Save More
EW-32661-19 Mfr # GFCS-011496
COMPARE
15 1/4" Compression Stainless Steel, Viton
$1,289.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-30 LPM, N2/Air, Stainless Steel Body
Buy More Save More
EW-32661-20 Mfr # GFCS-010064
COMPARE
30 1/4" Compression Stainless Steel, Viton
$1,289.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-100 LPM, N2/Air, Stainless Steel Body
Buy More Save More
EW-32661-24 Mfr # GFCS-010066
COMPARE
100 3/8" Compression Stainless Steel, Viton
$1,458.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-1000 LPM, N2/Air, Aluminum Body
Buy More Save More
EW-32665-10 Mfr # GFCS-011292
COMPARE
1000 3/4" NPT(F) Anodized Aluminum, Brass, Viton
$2,209.00USD / Each
Aalborg Compact Gas Mass Flow Controller, 0-500 LPM, N2/Air, Stainless Steel Body
Buy More Save More
EW-32666-00 Mfr # GFCS-011100
COMPARE
500 1/2" Compression Stainless Steel, Viton
$2,428.00USD / Each
Showing 1 - 20 of 28