This product is now discontinued, View Related Products
Air-Powered Double-Diaphragm Pumps, Low-Flow
Manifold design with dual, rotating inlet and outlet ports allow up to four inlet/outlet configurations—pump to two dispensing points or mix equal parts of two fluids
  • Two-stage air valve—operation at low air flow rates and pressures
  • Optional pulse dampener minimizes pulsation and pressure drops
  • Optional service kits include replacement diaphragms, check balls, and O-rings of the diaphragm material.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Inlet Connection Max Air Inlet Pressure (PSI) Max Flow Rate (GPM)
Availability Pricing
COMPARE
100 4.5
Call For Price
COMPARE
100 4.5
Call For Price
COMPARE
100 4.5
Call For Price
COMPARE
100 4.5
Call For Price
COMPARE
100 5
Call For Price
COMPARE
100 5
Call For Price
COMPARE
100 5
Call For Price
Showing 1- 7 of 7