Air-Powered Polypropylene Pump Head Standard Centrifugal Pumps
Air-operated pump is ideal for intrinsically safe areas
  • Use pumps in chemical plants, organic compound transfer applications, and wastewater treatment centers
Order an air lubricator and an air filter/regulator under accessories to remove impurities in the air supply.
$664.50 - $1,837.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Wetted Materials
Availability Pricing
COMPARE
6.2 32 PP pump head, ceramic shaft, Viton® O-ring, PP-encapsulated magnet
$664.50USD / Each
COMPARE
10.5 39 PP pump head, ceramic shaft, Viton® O-ring, PP-encapsulated magnet
$723.00USD / Each
COMPARE
18 50 SS pump head, ceramic shaft, Viton® O-ring, PP-encapsulated magnet
$1,837.50USD / Each