Air-Powered Venturi Vacuum pumps
No moving parts ensures long life with low maintenance
  • Venturi pumps are air-powered—require no electrical power source or lubricant
  • Place these compact vacuum pumps close to application for fast response and minimal vacuum loss
These pumps take advantage of the venturi effect—as a stream of compressed air exits a constricted orifice the resulting pressure drop draws air through a vacuum port. Since they require no oil or electricity, the resulting vacuum (up to 29.5" Hg) is clean and efficient. With no moving parts or electrical hazards, these air-powered pumps are also intrinsically safe. The remarkably compact size of these pumps enables you to design vacuum systems with smaller, less expensive components and reduced maintenance costs. Each model (except 79165-40) includes a replaceable silencer that reduces noise levels up to 30% while offering minimal resistance to air flow.
Select the model of vacuum pump best suited to your specific application. Model 78165-20 includes a vacuum gauge which provides easy visual monitoring—helpful when troubleshooting. We recommend an air filter upstream of the pump to prolong service life. Filter should have a pore size of 50 µm or smaller.
$125.75 - $398.75USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Max Vacuum (in Hg) Wetted Parts
Availability Pricing
COMPARE
0.5 28 Aluminum, Brass, Buna N
$148.50USD / Each
COMPARE
1.2 28 Aluminum, Brass, Buna N
$208.50USD / Each
COMPARE
3.2 28 Aluminum, Brass, Buna N
$351.50USD / Each
COMPARE
1.2 29.5 Aluminum, Brass, Buna N
$398.75USD / Each
COMPARE
0.3 27 Aluminum
$125.75USD / Each