Almatec Air-driven Postive Displacement Pumps
German precision engineered AODD pumps to get your application up and flowing.
The heart of the AODD is the air-valve and thoughtful German engineering went into the development of the precision control PERSWING P® air valve and its Air Distribution System (ADS). Resulting in a metal-free, pneumatically pilot-operated PERSWING P® (ADS) that ensures accurate reversal of the main piston and offers low noise levels, minimal maintenance and lube-free operation (due to two moving parts).
Almatec plastic pumps feature a solid body construction made out of virgin grade PE or PTFE materials that support the forces generated within a reciprocating pump. The high static mass leads to maximum process fluid containment and eliminates the need for external metal parts that would otherwise be used. The mechanical machining of a solid plastic block is economical thanks to modern CNC technology, enabling tight tolerances and maximum value. Almatec also feature best-in-class diaphragms that provide a smooth profile that is uninterrupted by any seals, maximizing containment and clean-ability. The diaphragms are designed to simplify assembly, reduce air consumption and optimize performance.
$739.25 - $7,081.00USD / Each
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Almatec E08 AODD PTFE Housing PTFE Diaphragm PTFE Valve Balls 1/4" NPT EW-75980-00 Mfr # E08TTT
COMPARE
AODD Pump, PTFE Housing PTFE Diaphragm PTFE Valve Balls 1/4" NPT
$1,253.50USD / Each
Almatec E08 AODD, Conductive PTFE Housing, PTFE Diaphragm, PTFE Valve Balls, 1/4" NPT EW-75980-02 Mfr # E08UTT
COMPARE
AODD Pump, Conductive PTFE Housing, PTFE Diaphragm, PTFE Valve Balls, 1/4" NPT
$1,339.00USD / Each
Almatec E10 AODD, Conductive PTFE Housing, PTFE Diaphragm, PTFE Valve Balls, 3/8" NPT EW-75980-06 Mfr # E10UTT
COMPARE
AODD Pump, Conductive PTFE Housing, PTFE Diaphragm, PTFE Valve Balls, 3/8" NPT
$1,875.00USD / Each
Almatec E15 AODD, Polyethylene Housing, EPDM Diaphragm, EPDM Valve Balls, 1/2" NPT EW-75980-08 Mfr # E15EEE
COMPARE
AODD Pump, Polyethylene Housing, EPDM Diaphragm, EPDM Valve Balls, 1/2" NPT
$1,339.00USD / Each
Almatec E15 AODD, Polyethylene Housing, PTFE Diaphragm, PTFE Valve Balls, 1/2" NPT EW-75980-10 Mfr # E15ETT
COMPARE
AODD Pump, Polyethylene Housing, PTFE Diaphragm, PTFE Valve Balls, 1/2" NPT
$1,628.50USD / Each
Almatec E15 AODD, Conductive Polyethylene Housing, EPDM Diaphragm, EPDM Valve Balls, 1/2" NPT EW-75980-12 Mfr # E15FEE
COMPARE
AODD Pump, Conductive Polyethylene Housing, EPDM Diaphragm, EPDM Valve Balls, 1/2" NPT
$1,382.00USD / Each
Almatec E15 AODD, Conductive Polyethylene Housing, PTFE Diaphragm, PTFE Valve Balls, 1/2" NPT EW-75980-14 Mfr # E15FTT
COMPARE
AODD Pump, Conductive Polyethylene Housing, PTFE Diaphragm, PTFE Valve Balls, 1/2" NPT
$1,671.50USD / Each
Almatec E15 AODD, Conductive PTFE Housing, PTFE Diaphragm, PTFE Valve Balls, 1/2" NPT EW-75980-18 Mfr # E15UTT
COMPARE
AODD Pump, Conductive PTFE Housing, PTFE Diaphragm, PTFE Valve Balls, 1/2" NPT
$3,642.50USD / Each
Almatec E25 AODD, Polyethylene Housing, PTFE Diaphragm, PTFE Valve Balls, 1" NPT EW-75980-22 Mfr # E25ETT
COMPARE
AODD Pump, Polyethylene Housing, PTFE Diaphragm, PTFE Valve Balls, 1" NPT
$2,142.50USD / Each
Almatec E25 AODD, Conductive Polyethylene Housing, EPDM Diaphragm, EPDM Valve Balls, 1" NPT EW-75980-24 Mfr # E25FEE
COMPARE
AODD Pump, Conductive Polyethylene Housing, EPDM Diaphragm, EPDM Valve Balls, 1" NPT
$1,853.50USD / Each
Almatec E25 AODD, Conductive Polyethylene Housing, PTFE Diaphragm, PTFE Valve Balls, 1" NPT EW-75980-26 Mfr # E25FTT
COMPARE
AODD Pump, Conductive Polyethylene Housing, PTFE Diaphragm, PTFE Valve Balls, 1" NPT
$2,228.50USD / Each
Almatec E25 AODD, PTFE Housing, PTFE Diaphragm, PTFE Valve Balls, 1" NPT EW-75980-28 Mfr # E25TTT
COMPARE
AODD Pump, PTFE Housing, PTFE Diaphragm, PTFE Valve Balls, 1" NPT
$6,556.00USD / Each
Almatec E25 AODD, Conductive PTFE Housing, PTFE Diaphragm, PTFE Valve Balls, 1" NPT EW-75980-30 Mfr # E25UTT
COMPARE
AODD Pump, Conductive PTFE Housing, PTFE Diaphragm, PTFE Valve Balls, 1" NPT
$7,081.00USD / Each
Almatec FUTUR 20 AODD, max. 100-130°C High Temperature PTFE Housing, PTFE Diaphragms, PTFE Valve Balls, 1/2" NPT EW-76980-02 Mfr # Fut20T
COMPARE
AODD Pump, Max. 100-130°C High Temperature PTFE Housing, PTFE Diaphragms, PTFE Valve Balls, 1/2" NPT
$4,038.50USD / Each
Almatec FUTUR 50 AODD, max. 100-130°C High Temperature PTFE Housing, PTFE Diaphragms, PTFE Valve Balls, 1" NPT EW-76980-04 Mfr # Fut50T
COMPARE
AODD Pump, Max. 100-130°C High Temperature PTFE Housing, PTFE Diaphragms, PTFE Valve Balls, 1" NPT
$6,363.00USD / Each
Almatec FUTUR 50 AODD, max. 70°Abrasion Resistant Polyethylene Housing, PTFE Diaphragms, PTFE Valve Balls, 1" NPT EW-76980-12 Mfr # Fut50E
COMPARE
AODD Pump, Max. 70° Abrasion Resistant Polyethylene Housing, PTFE Diaphragms, PTFE Valve Balls, 1" NPT
$3,888.50USD / Each
Almatec CXM 10 Mid-Range Duty AODD,  Conductive Polyethylene, PTFE Diaphragms,PTFE Valve Balls, 3/8" NPT EW-77980-01 Mfr # CXM10FTT
COMPARE
Mid-Range Duty AODD Pump, Conductive Polyethylene, PTFE Diaphragms,PTFE Valve Balls, 3/8" NPT
$739.25USD / Each
Almatec CXM 20 Mid-Range Duty AODD,  Conductive Polyethylene, EPDM Diaphragms,EPDM Valve Balls, 1/2" NPT, 90°-inlet-outlet EW-77980-03 Mfr # CXM20FEE
COMPARE
Mid-Range Duty AODD Pump, Conductive Polyethylene, EPDM Diaphragms,EPDM Valve Balls, 1/2" NPT, 90°-inlet-outlet
$777.75USD / Each
Almatec CXM 20 Mid-Range Duty AODD,  Conductive Polyethylene, PTFE Diaphragms,PTFE Valve Balls, 1/2" NPT, 90°-inlet-outlet EW-77980-05 Mfr # CXM20FTT
COMPARE
Mid-Range Duty AODD Pump, Conductive Polyethylene, PTFE Diaphragms,PTFE Valve Balls, 1/2" NPT, 90°-inlet-outlet
$918.25USD / Each
Almatec CP 6 Laboratory AODD,  PTFE, PTFE Diaphragms,PTFE Check Valve, 1/2" BSP inlet, 1/4" BSP oulet EW-77980-20 Mfr # CP6T
COMPARE
Laboratory AODD Pump, PTFE, PTFE Diaphragms,PTFE Check Valve, 1/2" BSP inlet, 1/4" BSP oulet
$1,425.00USD / Each
Showing 1 - 20 of 30