Alnor Micromanometers
Premium features make it ideal for HVAC, environmental safeguards, commissioning, process control and system balancing
  • Measure differential and static pressure from -15 to +15 in. H2O (-3735 to +3735 Pa)
  • Calculates and displays velocity when using a Pitot tube
  • Calculates volumetric flow rate in duct from velocity and user-input duct size and shape
  • Records data points in duct traverse using sampling function
  • Data logging with time and date stamp
  • Programmable K factors
The Alnor® AXD620 Micromanometer is a rugged, compact, comprehensive micromanometer that measures pressure. It can be used with pitot tubes to measure velocity and calculate flow rates with user–input duct size and shape.
Ideal applications include HVAC commissioning and troubleshooting, testing and balancing, Pitot tube duct traverses, static pressure measurements and differential pressure measurements.
$1,111.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
TSI Alnor AXD620 Micromanometer System with Data Logging Software Kit EW-10375-37 Mfr # AXD620
COMPARE
15500
$1,111.00USD / Each