Aluminum/Stainless Steel Gear Flowmeters 1801
  • Affordable flowmeters with ±0.5% accuracy are now available for flow rates up to 60 GPM!
  • High-strength aluminum or stainless steel body withstands pressures up to 5000 psi
  • Choose models with pulse output, 4 to 20 mA signal output, or integral digital display
Precisely measured gear teeth deliver consistent ±0.5% accuracy even when measuring high-viscosity fluids—ideal for measuring the flow of oils, grease, fuels, solvents, and other nonabrasive lubricating liquids. These positive displacement flowmeters are solidly built to provide excellent dynamic response for use in high-pressure applications. Gears and bearings withstand bidirectional flow without damage. Two 6-mm mounting holes let you secure the meter on a base plate, panel, or manifold. Available with a high-strength aluminum or 303 SS body. Pulse Output Flowmeters provide a NPN sourcing square wave pulse that is proportional to the flow rate. The peak-to-peak voltage of this square wave is the supply voltage provided minus two volts. &
$1,688.00 - $5,337.00USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Output Wetted Materials
Availability Pricing
AW-Lake Aluminum flowmeter with pulse output, 2.0 GPM EW-32928-00 Mfr # JVA-20KG + HEF-1
COMPARE
2 Square wave and 10 to 24 VDC 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, Aluminum
$1,688.00USD / Each
AW-Lake Aluminum flowmeter with pulse output, 0.5 GPM EW-32928-01 Mfr # JVA-12KG + HEF-1
COMPARE
0.5 Square wave and 10 to 24 VDC 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, Aluminum
$1,895.00USD / Each
AW-Lake Aluminum flowmeter with pulse output, 20 GPM EW-32928-02 Mfr # JVA-60KG + HEF-1
COMPARE
20 Square wave and 10 to 24 VDC 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, Aluminum
$2,048.00USD / Each
AW-Lake Stainless steel flowmeter with pulse output, 2.0 GPM EW-32928-10 Mfr # JVM-20KG + HEF-1
COMPARE
2 Square wave and 10 to 24 VDC 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, 303 Stainless Steel
$2,070.00USD / Each
AW-Lake Stainless steel flowmeter with pulse output, 20 GPM EW-32928-12 Mfr # JVM-60KG + HEF-1
COMPARE
20 Square wave and 10 to 24 VDC 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, 303 Stainless Steel
$2,859.00USD / Each
AW-Lake Aluminum meter-mounted analog flowmeter with 4 to 20 mA output, 2.0 GPM EW-32928-20 Mfr # JVA-20KG + FIP 4HS
COMPARE
2 4 to 20 mA 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, Aluminum
$1,800.00USD / Each
AW-Lake Aluminum meter-mounted analog flowmeter with 4 to 20 mA output, 20 GPM EW-32928-22 Mfr # JVA-60KG + FIP 4HS
COMPARE
20 4 to 20 mA 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, Aluminum
$2,160.00USD / Each
AW-Lake Stainless steel meter-mounted analog flowmeter with 4 to 20 mA output, 2.0 GPM EW-32928-26 Mfr # JVM-20KG + FIP 4HS
COMPARE
2 4 to 20 mA 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, 303 Stainless Steel
$2,195.00USD / Each
AW-Lake Stainless steel meter-mounted analog flowmeter with 4 to 20 mA output, 20 GPM EW-32928-28 Mfr # JVM-60KG + FIP 4HS
COMPARE
20 4 to 20 mA 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, 303 Stainless Steel
$2,972.00USD / Each
AW-Lake Aluminum meter-mounted digital flowmeter with integral display, 20 GPM EW-32928-32 Mfr # JVA-60KG + RT-10A
COMPARE
20 None 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, Aluminum
$2,600.00USD / Each
AW-Lake Stainless steel meter-mounted digital flowmeter with integral display, 2.0 GPM EW-32928-36 Mfr # JVM-20KG + RT-EX10A
COMPARE
2 None 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, 303 Stainless Steel
$2,622.00USD / Each
AW-Lake Stainless steel meter-mounted digital flowmeter with integral display, 20 GPM EW-32928-38 Mfr # JVM-60KG + RT-EX10A
COMPARE
20 None 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, 303 Stainless Steel
$3,423.00USD / Each
AW-Lake Stainless steel meter-mounted digital flowmeter with integral display, 60 GPM EW-32928-40 Mfr # JVM-80KG + RT-EX10A
COMPARE
60 None 17-4 PH Stainless Steel, 440 Stainless Steel, PTFE, 303 Stainless Steel
$5,337.00USD / Each
Showing 1- 13 of 13