Cole-Parmer® Essentials Aluminum Weighing Dishes, Crimped Wall with Tab
Crimpled walled with side finger tab handle
  • Disposable
Crimped sides for rigidity, sturdy gauge for heavy duty use. Large tab for sample numbers, flat bottom to improve sample handling, sides are angled for easy seperation. No oil residue, no vinyl coating to create contamination problems. Smooth bottom and may be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical.
$25.00 - $225.00USD / Pkg of 100
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Depth (in) Diameter (mm)
Availability Pricing
COMPARE
50 5/8 57
$155.00USD / Case of 1000
COMPARE
20 1/2 43
$139.00USD / Case of 1000
COMPARE
60 5/8 70
$180.00USD / Case of 1000
COMPARE
8 3/8 28
$225.00USD / Case of 500
COMPARE
50 5/8 57
$25.00USD / Pkg of 100
COMPARE
20 1/2 43
$28.00USD / Pkg of 100
COMPARE
60 5/8 70
$25.30USD / Pkg of 100
Showing 1- 7 of 7