American Machine & Tool 316 Stainless Steel Self-Priming Centrifugal Pumps
Chamber in front of pump housing enables self-priming operation
  • Self-cleaning, semi-open impellers
These American Machine & Tool 316 stainless steel self-priming centrifugal pumps feature a continuous duty TEFC motor. Pumps are constructed of a 316 stainless steel impeller and carbon graphite and ceramic bearing. Models 70620-00, -02, -10. -12, -22, and -32 feature a Viton® o-ring and seal. Models 70620-72 and -80 feature a PTFE o-ring and seal.
$1,632.00 - $1,931.50USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Power (VAC)
Availability Pricing
American Machine & Tool Self-Priming 316SS Centrifugal Pump, 59 GPM; 230/460 VAC EW-70620-02 Mfr # 282F-98
COMPARE
59 70 230 / 460
$1,632.00USD / Each
American Machine & Tool Self-Priming 316SS Centrifugal Pump, 78 GPM; 115/230 VAC EW-70620-10 Mfr # 282C-98
COMPARE
78 78 115 / 230
$1,664.00USD / Each
American Machine & Tool Self-Priming Centrifugal Pump, 316 SS, 78 GPM; 230/460 VAC EW-70620-12 Mfr # 282K-98
COMPARE
78 78 230 / 460
$1,723.00USD / Each
American Machine & Tool Self-Priming Centrifugal Pump, 316 SS, 100 GPM, 2 hp; 230/460 VAC EW-70620-22 Mfr # 282B-98
COMPARE
100 95 230 / 460
$1,770.50USD / Each
American Machine & Tool Self-Priming Centrifugal Pump, 316 SS, 125 GPM; 230/460 VAC EW-70620-32 Mfr # 282M-98
COMPARE
125 85 230 / 460
$1,931.50USD / Each
American Machine & Tool Self-Priming Centrifugal Pump, 316 SS, 100 GPM, 1.5 hp; 230/460 VAC EW-70620-72 Mfr # 3893-98
COMPARE
100 51 230 / 460
$1,653.00USD / Each
American Machine & Tool Self-Priming Centrifugal Pump, 316 SS,129 GPM; 230/460 VAC EW-70620-80 Mfr # 3894-98
COMPARE
129 82 230 / 460
$1,744.00USD / Each
Showing 1- 7 of 7