Ametek IS Submersible Level Transmitters
Design provides for excellent linearity and repeatability, low hysteresis, and long-term stability
  • Solid-state semiconductor sensor for accuracy and reliability
  • Rugged, 316 stainless steel housing with excellent environmental protection
  • 2-wire 4 to 20 mA output
  • Vented to the atmosphere through the surface end of the cable
  • Reverse polarity and surge protected
  • Long-lasting service with virtually no maintenance
  • Meet CSA requirements for intrinsically safe operation in hazardous locations as designated by Class I, Div I, Groups A, B, C, and D and Class II, Groups E, F, and G
The Model 575 transmitter offers you the convenience of direct submergence in many types of liquids, especially wastewater, for quick, accurate, and reliable level measurement. Simply lower the transmitter into a vessel or well—all the electronics are mounted in a submersible 316 stainless steel (SS) housing. The removable, nonclogging snub nose end protects the sensing elements.
The transmitter indicates the level of liquid by continuously measuring hydrostatic pressure via its sensing element, an ion-implanted silicon semiconductor chip with integral Wheatstone Bridge circuit. Once the sensor measures the pressure, the data is transmitted by a 4 to 20 mA output signal. This design provides for excellent linearity and repeatability, low hysteresis, and long-term stability.
$768.00 - $1,916.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI)
Availability Pricing
Ametek 575PB0006RHS Submersible Level Transmitter 0-6 Psi/ 0-14 Ft. EW-68049-42 Mfr # 575-S-B-0006-R-L-S-14
COMPARE
6
$809.00USD / Each
Ametek 575P B 0015 N H S 0-35" Submersible Level Transmitter 0-15 Psi/ 0-35 Ft. SKU 6804944 EW-68049-44 Mfr # 575-S-B-0015-R-L-S-35
COMPARE
15
$1,144.00USD / Each
Ametek 575P B 0015 N H S 0-35" Submersible Level Transmitter 0-15 Psi/ 0-35 Ft. SKU 6804945 EW-68049-45 Mfr # 575SB0030NLS(100) 0-69FT WTR
COMPARE
$1,122.00USD / Each
Ametek 575P 0-60 PSI/ 0-138FT Submersible Level Transmitter 0-60 Psi/ 0-138 Ft. EW-68049-46 Mfr # 575-S-B-0060-R-L-S-138
COMPARE
60
$768.00USD / Each
Ametek 575P B 0015 N H S 0-35" Submersible Level Transmitter 0-200 Psi/ 0-460 Ft. EW-68049-49 Mfr # 575SB0200NLS(550) 0-460FT WTR
COMPARE
$1,916.00USD / Each