This product is now discontinued, View Related Products

Ampicillin Trihydrate

  • Assay - ≥90%
  • pH of a 1% Solution at 25°C - 3.5 to 6.0
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher BioReagents BP90225 Ampicillin Trihydrate (Off-white Powder) (25g) EW-88068-47 Mfr # BP90225
COMPARE
7177-48-2
Call For Price
Acros Organics AC612290250 Ampicillin trihydrate (25g) EW-88365-85 Mfr # AC612290250
COMPARE
7177-48-2
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat