Analytik Jena High Intensity Shortwave Ultraviolet Lamps
This high-intensity, ultraviolet lamp has a 2" x 6" filter area (5000 hour filter life) and a high-intensity grid. Lamp emits 254 nm light with an intensity of 2400 µW/cm² at a distance of 5". A 3-ft cord connects lamp to base; an 8-ft, three-wire cord connects base to outlet. The 115 VAC model comes with a plug; the 230 VAC model has stripped ends.
$1,790.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (watts)
Availability Pricing
Analytik Jena High-intensity, shortwave, grid UV lamp; 115 VAC/60 Hz EW-09813-00 Mfr # 94-0012-05
COMPARE
$1,790.00USD / Each