Analytik Jena Versatile Handheld Ultraviolet Lamps

Switch between light sources or wavelength at the touch of a button
  • Triple-wavelength model available for excellent versatility
$921.00 - $933.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Wavelength
Availability Pricing
Analytik Jena Handheld UV Lamp, 254/302/365 nm Wavelength; 115 VAC, 60 Hz EW-97607-30 Mfr # 95-0252-01
COMPARE
60 115 254/302/365 nm
$933.00USD / Each
Analytik Jena Handheld UV Lamp, 254/302/365 nm Wavelength; 230 VAC, 60 Hz EW-97607-35 Mfr # 95-0252-02
COMPARE
50 230 254/302/365 nm
$921.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat