Aprotinin

  • Description - Clear, Colorless Solution
  • Activity - ≈ 10,000 KIU/mL
  • λ Max - 277 ± 3nm
  • Sodium Chloride - ≈ 9mg/mL
  • Benzyl Alcohol - ≈ 9mg/mL
  • pH - 5.7-6.4
$97.00 - $1,188.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher BioReagents BP250310 Aprotinin (From Bovine Lung) (10 mL) CAS 9087-70-1 EW-88059-70 Mfr # BP250310
COMPARE
9087-70-1 Analytical Reagent
$97.00USD / Each
Fisher BioReagents BP250340 Aprotinin (From Bovine Lung) (40 mL) CAS 9087-70-1 EW-88059-71 Mfr # BP250340
COMPARE
9087-70-1 Analytical Reagent
$1,188.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat