Argon Gas Mass Flowmeters with Metal bodies
Precisely measure flow rates and totals
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These flowmeters feature an advanced straight-tube sensor that ensures accurate and repeatable
results. Gas flow measurements are unaffected by moderate temperature and pressure variations
at the inlet. The meter also allows a four-point calibration across the flow range to improve meter
linearity. Output data from the meter can be sent via a 0 to 5 VDC or 4 to 20 mA signal; an analogto-
RS converter is available for data collection and analysis on your PC.
All meters include a detachable 3 1/2-digit LCD that can be tilted up to 90° for easy reading.
Remotely mount the display up to three feet using extension cable 32662-70. The optional totalizer
32650-22 displays accumulated flow. The meter is protected from polarity reversal or short circuit
by a built-in resettable fuse. Aluminum models have wetted materials of anodized aluminum,
brass, and Viton®; stainless steel (SS) models have wetted materials of SS and Viton.
Accuracy:
These precalibrated flow controllers operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77°F (15 to 25°C) while maintaining the stated percent full-scale accuracy and linearity. When operating beyond 5 to 60 psi, add ±0.01%/psi full-scale; if operating beyond 59 to 77°F (15 to 25°C), add ±0.15%/°C full-scale.
$650.00 - $1,987.00USD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Process Connection Wetted Materials
Availability Pricing
Aalborg Thermal Gas Mass Flowmeter, to 10 sccm Ar, Aluminum body w/ display
Buy More Save More
EW-32657-00 Mfr # GFMS-010179
COMPARE
0.01 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 100 sccm Ar, Aluminum body w/ display
Buy More Save More
EW-32657-06 Mfr # GFMS-010182
COMPARE
0.1 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 200 sccm Ar, Aluminum body w/ display
Buy More Save More
EW-32657-08 Mfr # GFMS-010183
COMPARE
0.2 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 1 L/min Ar, Aluminum body w/ display
Buy More Save More
EW-32657-12 Mfr # GFMS-010185
COMPARE
1 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 2 L/min Ar, Aluminum body w/ display
Buy More Save More
EW-32657-14 Mfr # GFMS-010186
COMPARE
2 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 5 L/min Ar, Aluminum body w/ display
Buy More Save More
EW-32657-16 Mfr # GFMS-010187
COMPARE
5 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$650.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 30 L/min Ar, Aluminum body w/ display
Buy More Save More
EW-32657-34 Mfr # GFMS-010191
COMPARE
30 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$814.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 100 L/min Ar, Aluminum body w/ display
Buy More Save More
EW-32657-42 Mfr # GFMS-011236
COMPARE
100 3/8" Compression Fittings Anodized Aluminum, Brass, Viton
$1,000.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 500 L/min Ar, Aluminum body w/ display
Buy More Save More
EW-32657-44 Mfr # GFMS-011014
COMPARE
500 1/2" Compression Fittings Anodized Aluminum, Brass, Viton
$1,263.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 10 sccm Ar, 316SS Body
Buy More Save More
EW-32657-50 Mfr # GFMS-010194
COMPARE
0.01 1/4" Compression Fittings 316 Stainless Steel, Viton
$984.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 50 sccm Ar, 316SS Body
Buy More Save More
EW-32657-54 Mfr # GFMS-010196
COMPARE
0.05 1/4" Compression Fittings 316 Stainless Steel, Viton
$984.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 100 sccm Ar, 316SS Body
Buy More Save More
EW-32657-56 Mfr # GFMS-010197
COMPARE
0.1 1/4" Compression Fittings 316 Stainless Steel, Viton
$984.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 2 L/min Ar, 316SS Body
Buy More Save More
EW-32657-64 Mfr # GFMS-010201
COMPARE
2 1/4" Compression Fittings 316 Stainless Steel, Viton
$984.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 15 L/min Ar, 316SS Body
Buy More Save More
EW-32657-68 Mfr # GFMS-011387
COMPARE
15 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,222.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 30 L/min Ar, 316SS Body
Buy More Save More
EW-32657-84 Mfr # GFMS-010204
COMPARE
30 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,222.00USD / Each
Aalborg GFMS Thermal Gas Mass Flowmeter with Display, 50 L/min Ar, 316SS Body
Buy More Save More
EW-32657-86 Mfr # GFMS-010205
COMPARE
50 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,222.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 200 L/min Ar, 316SS body w/ display
Buy More Save More
EW-32659-06 Mfr # GFMS-011134
COMPARE
200 3/8" Compression Fittings 316 Stainless Steel, Viton
$1,656.00USD / Each
Aalborg Thermal Gas Mass Flowmeter, to 1000 L/min Ar, 316SS body w/ display
Buy More Save More
EW-32659-26 Mfr # GFMS-012406
COMPARE
1000 3/4" NPT(F) Fittings 316 Stainless Steel, Viton
$1,987.00USD / Each
Showing 1- 18 of 18