Argos Technologies® Attached Cap 5 mL Microcentrifuge Tubes
Easy-to-open cap allows for single-handed operation
  • Safely centrifuge up to 5000 g
  • Certified RNase-, DNase-, and pyrogen-free
The graduated polypropylene (PP) microcentrifuge tubes offer an alternative to 15-mL conical tubes for applications involving volumes less than 5 mL. The smaller tube size allows only the pipette tip to be inserted, reducing risk of contamination.
Easy-to-open attached cap eliminates hand fatigue and maintains sample integrity. Use frosted marking area for tagging and identification. The tubes can withstand autoclaving to 248°F (120°C) for 15 minutes at 15 psi, and are suitable for freezing to –130°F (–90°C).
$59.00 - $65.00USD / Pkg of 200
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Microcentrifuge Tubes, 5 mL, Non-Sterile, Clear; 200/Pk
$59.00USD / Pkg of 200
COMPARE
Microcentrifuge Tubes, 5 mL, Sterile, Clear; 20/Bag, 10 Bag/Cs
$65.00USD / Case of 200
COMPARE
Microcentrifuge Tubes, 5 mL, Non-Sterile, Blue; 200/Pk
$59.00USD / Pkg of 200
COMPARE
Microcentrifuge Tubes, 5 mL, Non-Sterile, Yellow; 200/Pk
$59.00USD / Pkg of 200
COMPARE
Microcentrifuge Tubes, 5 mL, Non-Sterile, Green; 200/Pk
$59.00USD / Pkg of 200
COMPARE
Microcentrifuge Tubes, 5 mL, Non-Sterile, Pink; 200/Pk
$59.00USD / Pkg of 200
COMPARE
Microcentrifuge Tubes, 5 mL, Non-Sterile, Black; 200/Pk
$60.00USD / Pkg of 200
Showing 1- 7 of 7