Argos Technologies® BackSpring Microcentrifuge Tubes
Eliminate sore thumbs and repetitive stress on your hand from opening tubes
  • RNase, DNase, and pyrogen free
  • Boil-proof and withstands freezing up to –90°C
  • Easy-to-read graduations with frosted writing surface
  • Can be spun up to 30,000 g
  • Easy puncture top
Reduce the stress on your hand due to repeatedly opening and closing microcentrifuge tubes. This unique 1.5-mL tube lets you open it from the back with just one hand, versus the traditional front-open style. Pivot design allows the tube to snap open when pressure is applied to the back of the cap — enabling quick, comfortable opening and closing of the lid. Withstands temperatures from -90° to 121°C. Autoclavable.
$16.80 - $20.50USD / Pkg of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Sterile Tube Capacity (mL) Description
Availability Pricing
COMPARE
No 1.5 BackSpring Microcentrifuge Tube with Integrated Cap, 1.5 mL; 500/Pk
$16.80USD / Pkg of 500
COMPARE
Yes 1.5 BackSpring Microcentrifuge Tube with Integrated Cap, 1.5 mL, Sterile; 500/Pk
$20.50USD / Pkg of 500