Argos Technologies® Flexifuge Centrifuge Rotors
Versatile rotor options suit many standard lab needs
  • Quickly and easily interchange rotors
  • Constructed of durable polycarbonate
Replacement rotors and for the Argos Technologies® Flexifuge Centrifuge, models 22999-50 through -52.
$40.25 - $56.75USD / Pkg of 8
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Tubes Accepted Rotor Type Tube Sizes Accepted
Availability Pricing
COMPARE
8 Fixed-angle 1.5 or 2.0 mL microtubes
$56.00USD / Each
COMPARE
16 Fixed-angle Standard PCR tubes
$56.25USD / Each
COMPARE
4 Fixed-angle 5 mL
$56.75USD / Each
COMPARE
1 Fixed-angle 0.5 mL microtubes
$52.00USD / Pkg of 8
COMPARE
4 Fixed-angle 5 mL CapSafe
$40.25USD / Each