Argos Technologies® Flexifuge Microcentrifuge
Change color of illuminated LED light ring to personalize sample spinning
  • Quick centrifugation in a tiny footprint
  • Versatile rotor options suit many standard lab needs including 5-mL microtubes
  • Operate in continuous, pulsed, or timed mode
This compact versatile benchtop centrifuge is ideal for quick two-speed spin downs. Three rotors accommodate PCR, 0.5-mL, 1.5-mL, and 5-mL tubes. Operate the centrifuge in continuous mode, pulsed for a short burst of rapid centrifugation, or timed to complete the spin in 1 to 15 minutes.
Regardless of setting, rotor will stop once the lid release button is pushed.
An outer ring of colored LED lights illuminate when the unit is active, and can be easily viewed through the transparent lid even in a dark room. The lights can be turned off or the color changed to red, green, or blue by a switch underneath the unit.
$498.00 - $537.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrical Plug Type Power (VAC)
Availability Pricing
COMPARE
US 120
$498.00USD / Each
COMPARE
Europe 220
$537.00USD / Each