Cole-Parmer® Essentials Pipette Pumps
Economical choice for accurate dispensing
  • Resists acids and alkalis
  • Color coded for easy identification
  • Unit disassembles for easy cleaning
Manual pipette pumps feature a thumbwheel for precision filling or dispensing of liquids. Insert glass or plastic pipettes into secure tapered collar and quickly dispense liquid by depressing white side bar or plunger. Operate easily with one hand.
$19.00 - $22.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Manual Pipette Pump, 10 mL, Green
$19.00USD / Each
COMPARE
Manual Pipette Pump, 25 mL, Red
$22.00USD / Each