Cole-Parmer® Essentials Upright Freezer Drawer Racks for 96 Deep-Well Microtiter Plates or Boxes w/1 3/4" Max Height
Drawers provide quick storage and retrieval of plates and microtube boxes
  • Easily access fewer microtiter plates and boxes at a time, protecting other samples in the rack from warming up
  • Durable corrosion-resistant stainless steel construction for long-term use
Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer.
$164.00 - $543.00USD / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
3 x 3 Array 6 1/16 10 15/16
$164.00USD / Each
COMPARE
3 x 4 Array 8 1/8 10 15/16
$206.25USD / Each
COMPARE
4 x 3 Array 6 1/16 14 1/2
$199.00USD / Each
COMPARE
4 x 4 Array 8 1/8 14 1/2
$301.00USD / Each
COMPARE
4 x 5 Array 10 1/8 14 1/2
$303.00USD / Each
COMPARE
4 x 6 Array 12 1/16 14 1/2
$325.50USD / Each
COMPARE
5 x 3 Array 6 1/16 18
$235.00USD / Each
COMPARE
5 x 4 Array 8 1/8 18
$305.00USD / Each
COMPARE
5 x 5 Array 10 1/8 18
$368.75USD / Each
COMPARE
5 x 6 Array 12 1/16 18
$402.75USD / Each
COMPARE
6 x 3 Array 6 1/16 21 1/2
$271.00USD / Each
COMPARE
6 x 4 Array 8 1/8 21 1/2
$330.75USD / Each
COMPARE
6 x 5 Array 10 1/8 21 1/2
$418.25USD / Each
COMPARE
6 x 6 Array 12 1/16 21 1/2
$461.50USD / Each
COMPARE
6 x 7 Array 14 1/16 21 1/2
$543.00USD / Each
COMPARE
7 x 4 Array 8 1/8 25
$391.50USD / Each
COMPARE
7 x 5 Array 10 1/8 25
$466.75USD / Each
Showing 1- 17 of 17