ResinTech Drop-In Style Carbon Block and Radial Flow Cartridges
Eliminate harmful contaminants and improve drinking water
  • Higher capacity cartridges with more media extend cartridge life
  • Designed to fit point-of-entry housings and high flow applications
  • Ensure quality control of each cartridge with complete traceability
  • Made in the USA!
These carbon block and radial flow cartridges are an inexpensive and effective way to remove chemicals and contaminants commonly found in tap water. These are ideal for taste and odor control as part of a water treatment system.
Each cartridge is wrapped with a polypropylene outer layer and netting to prevent premature clogging of the carbon block.
Choose from a wide range of filter cartridge options depending on the specific contaminants that you need to eliminate.
$71.10 - $139.90USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description Capacity (Gal) Compatible with
Availability Pricing
ResinTech RC-10-1052-BB Radial Flow Carbon Filter Cartridge, 10" Big Blue EW-99285-71 Mfr # RC-10-1052-BB
COMPARE
Radial Flow Granular Activated Carbon Filter, 4.5\" x 10\" 120000 10\" Standard Housing
$71.10USD / Each
ResinTech RC-20-1052-BB Radial Flow Carbon Filter Cartridge, 20" Big Blue EW-99285-72 Mfr # RC-20-1052-BB
COMPARE
Radial Flow Granular Activated Carbon Filter, 4.5\" x 20\" 240000 20\" Standard Housing
$139.90USD / Each