ARIES FilterWorks Phoenix Ultra High-Purity Water Systems

Obtain Type I, 18.2 MΩ water—on demand!
  • Save money—best cost-of-ownership on the market
  • Save time—2.5 LPM flowrate and quick change of consumables
  • Built-in leak detection protects your investment
  • Full-color touch-screen display with intuitive interface
The Phoenix systems use a staged purification process that includes high-purity ion exchange resins to remove dissolved minerals, and internal recirculation to further maintain water purity. Multiple sensors continuously monitor the system and final water quality to ensure trouble-free operation.

As water exits the system, a final filter removes particulates and bacteria to reach Type I water specifications. Additional options are available for special applications to include 185/254 nm UV, and or ultrafiltration (UF).

The Phoenix system is pre-configured into four application specific models with benchtop or remote dispesing options:
Phoenix Academy (models 99285-31 and -32): basic chemistry, academic, IC, and buffers
Phoenix Citation (models 99285-35 and -36 with UV): HPLC, GC-MS, and trace organics
Phoenix Biochem (models 99285-33 and -34 with UF): life science, cell culture, and microbiology
Phoenix Genome (models 99285-37 and -38 with UV and UF): DNA Sequencing, PCR, and electrophoresis

A complete package requires a system, conditioning cartridge, polishing cartridge, and final filter.
1. Choose base system
2. Choose conditioning cartridge 99285-45 for RO water feed, or 99285-46 for SDI/tap water feed
3. Choose polishing cartridge 99285-43 for standard results, or 99285-44 for additional organics removal
4. Choose final filter 99285-27 for up to 0.2µm pariculate removal, or 99285-47 to also include endotoxin removal

Options (must be ordered with unit, factory installation required):
Item 99285-39 - adds in flow totalizer function, viewable on display
Item 99285-40 - adds in a direct feed port
Item 99285-41 - adds in a recirculating dispensing gun (for units without a remote dispense function)
Item 99285-42 - adds in a wall mounting bracket to clear up bench space
$6,946.00 - $9,596.00USD / Each

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description With Ultrafilter With UV Lamp
Availability Pricing
Aries Filterworks Phoenix Academy Benchtop or Wall Mount Base System, 120V/60Hz EW-99285-31 Mfr # PHX-AD-S-1
COMPARE
Academy Benchtop or Wall Mount Base System, 120V/60Hz No No
$6,946.00USD / Each
Aries Filterworks Phoenix Academy Remote Dispense Base System 120V/60Hz EW-99285-32 Mfr # PHX-AD-R-1
COMPARE
Academy Remote Dispense Base System 120V/60Hz No No
$8,056.00USD / Each
Aries Filterworks Phoenix Biochem Benchtop System with Ultrafiltration module, 120V/60Hz EW-99285-33 Mfr # PHX-BC-S-1
COMPARE
Biochem Benchtop System with Ultrafiltration module, 120V/60Hz Yes No
$7,726.00USD / Each
Aries Filterworks Phoenix Biochem Remote Dispense System with Ultrafiltration module, 120V/60Hz EW-99285-34 Mfr # PHX-BC-R-1
COMPARE
Biochem Remote Dispense System with Ultrafiltration module, 120V/60Hz Yes No
$8,838.00USD / Each
Aries Filterworks Phoenix Citation Benchtop System with UV Oxidation Lamp, 120V/60Hz EW-99285-35 Mfr # PHX-CT-S-1
COMPARE
Citation Benchtop System with UV Oxidation Lamp, 120V/60Hz No Yes
$7,872.00USD / Each
Aries Filterworks Phoenix Citation Remote Dispense System with UV Oxidation Lamp 120V/60Hz EW-99285-36 Mfr # PHX-CT-R-1
COMPARE
Citation Remote Dispense System with UV Oxidation Lamp 120V/60Hz No Yes
$8,984.00USD / Each
Aries Filterworks Phoenix Genome Benchtop with UV and UF, 120V/60Hz EW-99285-37 Mfr # PHX-GN-S-1
COMPARE
Genome Benchtop with UV and UF, 120V/60Hz Yes Yes
$8,484.00USD / Each
Aries Filterworks Phoenix Genome Remote Dispense Configuration with UV and UF, 120V/60Hz EW-99285-38 Mfr # PHX-GN-R-1
COMPARE
Genome Remote Dispense Configuration with UV and UF, 120V/60Hz Yes Yes
$9,596.00USD / Each
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat