ASCO 068 Series Flapper Isolation Valves
Hermetic separation from control mechanisms prevents fluid contamination
  • High maximum pressure rating of 116 psi (8 bar)
  • Uses only 4 watts at ambient conditions
Designed to control the flow of neutral or aggressive fluids, this ASCO 068 series of pad-mounted valves is designed for complete separation of fluid and solenoid while allowing for easy flush and self-draining capability. Flapper mechanism features a fast response time and very precise volume control. Additionally, valve has no pumping or sticking effect, and is tested to over 10 million cycles for long-life use. Common applications include chromatography, sample preparations, chemical sterilization, and water quality monitoring.
Flapper isolation valves are designed for use in vacuum to positive pressure applications. Choose from FFKM, EPDM, and FKM seal materials that best suit your fluids'' compatibility. Body is PEEK. Port type of valve is a pad mount to blanking plate (not included).
Two-way normally open and three-way universal operation, as well as different port type valves are also available—contact an Application Specialist.
$119.90 - $254.30USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Cv Housing Material Orifice (in)
Availability Pricing
Asco R068212S0100F1 Pad-Mount Solenoid Valve, NC, 0.8 mm Orifice; PEEK Body, FFKM Diaphragm; 24VDC EW-98125-00 Mfr # R068212S0100F1
COMPARE
0.017 PEEK 0.031
$254.30USD / Each
Asco R068212S0V00F1 Pad-Mount Solenoid Valve, NC, 0.8 mm Orifice; PEEK Body, EPDM Diaphragm; 24VDC EW-98125-02 Mfr # R068212S0V00F1
COMPARE
0.017 PEEK 0.031
$123.10USD / Each
Asco R068212S0E00F1 Pad-Mount Solenoid Valve, NC, 0.8 mm Orifice; PEEK Body, FKM Diaphragm; 24VDC EW-98125-04 Mfr # R068212S0E00F1
COMPARE
0.017 PEEK 0.031
$119.90USD / Each
Asco R068214S0100F1 Pad-Mount Solenoid Valve, NC, 1.2 mm Orifice; PEEK Body, FFKM Diaphragm; 24VDC EW-98125-06 Mfr # R068214S0100F1
COMPARE
0.042 PEEK 0.047
$254.30USD / Each
Asco R068214S0V00F1 Pad-Mount Solenoid Valve, NC, 1.2 mm Orifice; PEEK Body, EPDM Diaphragm; 24VDC EW-98125-08 Mfr # R068214S0V00F1
COMPARE
0.042 PEEK 0.047
$123.10USD / Each
Asco R068214S0E00F1 Pad-Mount Solenoid Valve, NC, 1.2 mm Orifice; PEEK Body, FKM Diaphragm; 24VDC EW-98125-10 Mfr # R068214S0E00F1
COMPARE
0.042 PEEK 0.047
$119.90USD / Each
Asco R068216S0100F1 Pad-Mount Solenoid Valve, NC, 1.6 mm Orifice; PEEK Body, FFKM Diaphragm; 24VDC EW-98125-12 Mfr # R068216S0100F1
COMPARE
0.063 PEEK 0.063
$254.30USD / Each
Asco R068216S0V00F1 Pad-Mount Solenoid Valve, NC, 1.6 mm Orifice; PEEK Body, EPDM Diaphragm; 24VDC EW-98125-14 Mfr # R068216S0V00F1
COMPARE
0.063 PEEK 0.063
$123.10USD / Each
Asco R068216S0E00F1 Pad-Mount Solenoid Valve, NC, 1.6 mm Orifice; PEEK Body, FKM Diaphragm; 24VDC EW-98125-16 Mfr # R068216S0E00F1
COMPARE
0.063 PEEK 0.063
$119.90USD / Each
Showing 1- 9 of 9