ASCO Series 212 Water Conditioning and Purification Valves

NSF-certified valves provide control in water conditioning and purification systems
  • Approved for NSF 42, 61, and 169 standards
  • Lightweight PPE composite body valve with NPT(M) connections
Designed for commercial and industrial markets which use membrane-based reverse osmosis systems such as food and beverage industries. Series 212 pilot-operated solenoid valves are designed for your high pressure and temperature processes, withstanding up to 150 psi (10 bar) and 180°F (82°C).
$166.90 - $310.10USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cv Orifice (in) Port Size (NPT(F))
Availability Pricing
Asco 8212A001S0100F0 Solenoid Valve, NSF, NC; 3/8 NPT(F), 5/8 Orifice; PPE Body, EPDM Seal; 120VDC EW-98125-18 Mfr # 8212A001S0100F0
COMPARE
3 0.75 0.375
$166.90USD / Each
Asco 8212A019S0100F0 Solenoid Valve, NSF, NC; 1/2 NPT(F), 3/4 Orifice; PPE Body, EPDM Seal; 120VDC EW-98125-20 Mfr # 8212A019S0100F0
COMPARE
4.5 0.75 0.5
$166.90USD / Each
Asco 8212A039S0100F0 Solenoid Valve, NSF, NC; 3/4 NPT(F), 3/4 Orifice; PPE Body, EPDM Seal; 120VDC EW-98125-22 Mfr # 8212A039S0100F0
COMPARE
7 0.75 0.75
$203.00USD / Each
Asco 8212A055S0100F0 Solenoid Valve, NSF, NC; 1 NPT(F), 1 Orifice; PPE Body, EPDM Seal; 120VDC EW-98125-24 Mfr # 8212A055S0100F0
COMPARE
13 1 1
$310.10USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat