Asco Solenoid-Operated 2- and 3-Way Pinch Valves
Simple tubing replacement
These pinch valves are excellent for isolation-type applications as the only wetted component is the tubing. They feature removable coils with DIN-style electrical connectors and a panel mount bracket is included. Ideal for precise fluid handling applications, these valves have a 20 msec response time.
$174.00 - $243.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Pinch Valve Tube Size Power (VDC)
Availability Pricing
Asco SCH284A005- 12VDC Two-way normally closed pinch valve, 12 VDC, 1/8" x 3/16" EW-01340-54 Mfr # SCH284A005- 12VDC
COMPARE
Two-Way, Normally-Closed 1/8" ID x 3/16" OD 12
$174.00USD / Each
Asco SCH284A005- 24VDC Two-way normally closed pinch valve, 24 VDC, 1/8" x 3/16" EW-01340-55 Mfr # SCH284A005- 24VDC
COMPARE
Two-Way, Normally-Closed 1/8" ID x 3/16" OD 24
$177.00USD / Each
Asco SCH284B007- 12VDC Two-way normally closed pinch valve, 12 VDC, 1/4" x 3/8" EW-01340-56 Mfr # SCH284B007- 12VDC
COMPARE
Two-Way, Normally-Closed 1/4" ID x 3/8 OD 12
$243.00USD / Each
Asco SCH284B007- 24VDC Two-way normally closed pinch valve, 24 VDC, 1/4" x 3/8" EW-01340-57 Mfr # SCH284B007- 24VDC
COMPARE
Two-Way, Normally-Closed 1/4" ID x 3/8" OD 24
$235.00USD / Each
Asco SCH284A013- 12VDC Two-way normally open pinch valve, 12 VDC, 1/8" x 3/16" EW-01340-60 Mfr # SCH284A013- 12VDC
COMPARE
Two-Way, Normally-Open 1/8" ID x 3/16" OD 12
$174.00USD / Each
Asco SCH284A013- 24VDC Two-way normally open pinch valve, 24 VDC, 1/8" x 3/16" EW-01340-61 Mfr # SCH284A013- 24VDC
COMPARE
Two-Way, Normally-Open 1/8" ID x 3/16" OD 24
$174.00USD / Each
Asco SCH284B015- 12VDC Two-way normally open pinch valve, 12 VDC, 1/4" x 3/8" EW-01340-62 Mfr # SCH284B015- 12VDC
COMPARE
Two-Way, Normally-Open 1/4" ID x 3/8" OD 12
$236.00USD / Each
Asco SCH284B015- 24VDC Two-way normally open pinch valve, 24 VDC, 1/4" x 3/8" EW-01340-63 Mfr # SCH284B015- 24VDC
COMPARE
Two-Way, Normally-Open 1/4" ID x 3/8" OD 24
$236.00USD / Each
Asco SCH384A005- 12VDC 3-way pinch valve, 12 VDC, 1/8" x 3/16" (Solenoid Electric Valves (actuated)) EW-01340-66 Mfr # SCH384A005- 12VDC
COMPARE
Three-Way, One Normally-Open, One Normally-Closed 1/8" ID x 3/16" OD 12
$191.00USD / Each
Asco SCH384A005-24VDC 3-way pinch valve, 24 VDC, 1/8" x 3/16" (0.125 in ID x 0.1875 in OD) EW-01340-67 Mfr # SCH384A005-24VDC
COMPARE
Three-Way, One Normally-Open, One Normally-Closed 1/8" ID x 3/16" OD 24
$191.00USD / Each
Showing 1- 10 of 10
Need Help with this product?
Call Us At1-847-549-7600
Live chat