ASCO Subminiature Solenoid Valves
Stainless Steel construction
  • Two-way normal closed operation
  • Install in any position
  • Available with manual operator
$88.00 - $101.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description Cv Max Differential Pressure
Availability Pricing
Asco U8256A103E 24VDC Subminiature Solenoid Valve 1/8" NPT(F), 3/32" Orfice, 24VDC EW-01340-79 Mfr # U8256A103E 24VDC
COMPARE
Subminiature Solenoid Valve 1/8" NPT(F), 3/32" Orfice, 24VDC 0.15 180 PSI Water
$88.00USD / Each
Asco U8256A104E 120VAC Subminiature Solenoid Valve 1/8" NPT(F), 7/64" Orfice, 120VAC EW-01340-81 Mfr # U8256A104E 120VAC
COMPARE
Subminiature Solenoid Valve 1/8" NPT(F), 7/64" Orfice, 120VAC 0.17 126 PSI Water
$88.00USD / Each
Asco U8256A107E 120VAC Subminiature Solenoid Valve 1/4" NPT(F),3/32" Orfice, 120VAC EW-01340-85 Mfr # U8256A107E 120VAC
COMPARE
Subminiature Solenoid Valve 1/4" NPT(F),3/32" Orfice, 120VAC 0.15 200 PSI Water
$101.00USD / Each
Asco U8256A107E 24VDC Subminiature Solenoid Valve 1/4" NPT(F),3/32" Orfice, 24VDC EW-01340-87 Mfr # U8256A107E 24VDC
COMPARE
Subminiature Solenoid Valve 1/4" NPT(F),3/32" Orfice, 24VDC 0.15 200 PSI Water
$101.00USD / Each
Asco U8256A108E 120VAC Subminiature Solenoid Valve 1/4" NPT(F), 7/64" Orfice, 120VAC EW-01340-89 Mfr # U8256A108E 120VAC
COMPARE
Subminiature Solenoid Valve 1/4" NPT(F), 7/64" Orfice, 120VAC 0.17 150 PSI Water
$101.00USD / Each
Asco U8256A108E 24VDC Subminiature Solenoid Valve 1/4" NPT(F), 7/64" Orfice, 24VDC EW-01340-91 Mfr # U8256A108E 24VDC
COMPARE
Subminiature Solenoid Valve 1/4" NPT(F), 7/64" Orfice, 24VDC 0.17 150 PSI Water
$101.00USD / Each
Showing 1- 6 of 6
Need Help with this product?
Call Us At1-847-549-7600
Live chat