ASCO Two- and Three-Way Miniature Solenoid Valves
Corrosion-resistant materials of construction
  • Low power consumption
  • Manifold mount construction allows easy assembly
  • Ports are #10-32 UNF
Universal-operation, three-way valves allow you to introduce pressure at any port for gas or chemical mixing into the third port.
$66.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Cv Inlet Port Orifice (in)
Availability Pricing
Asco 411M1112HV Solenoid Valve, Two-Way Normally Closed, 12V, 0.65 Watts, 100 psi EW-98515-22 Mfr # 411M1112HV
COMPARE
0.013 #10-32 UNF 0.025
$66.00USD / Each
Asco 411M2112HV Solenoid Valve, Two-Way Normally Closed, 12V, 2 Watts, 100 psi EW-98515-24 Mfr # 411M2112HV
COMPARE
0.038 #10-32 UNF 0.055
$66.00USD / Each
Asco 411M3112HV Solenoid Valve, Two-Way Normally Closed, 12V, 2 Watts, 30 psi EW-98515-26 Mfr # 411M3112HV
COMPARE
0.07 #10-32 UNF 0.08
$66.00USD / Each
Asco 411M1124HV Solenoid Valve, Two-Way Normally Closed, 24V, 0.65 Watts, 100 psi EW-98515-28 Mfr # 411M1124HV
COMPARE
0.013 #10-32 UNF 0.025
$66.00USD / Each
Asco 411M2124HV Solenoid Valve, Two-Way Normally Closed, 24V, 2 Watts, 100 psi EW-98515-30 Mfr # 411M2124HV
COMPARE
0.038 #10-32 UNF 0.055
$66.00USD / Each
Asco 411M3124HV Solenoid Valve, Two-Way Normally Closed, 24V, 2 Watts, 30 psi EW-98515-32 Mfr # 411M3124HV
COMPARE
0.07 #10-32 UNF 0.08
$66.00USD / Each
Asco 411M1312HV Solenoid Valve, Three-Way Universal Operation, 12V, 0.65 Watts, 100 psi EW-98515-34 Mfr # 411M1312HV
COMPARE
0.013/0.008 #10-32 UNF 0.025/0.025
$66.00USD / Each
Asco 411M2312HV Solenoid Valve, Three-Way Universal Operation, 12V, 2 Watts, 50 psi EW-98515-36 Mfr # 411M2312HV
COMPARE
0.038/0.03 #10-32 UNF 0.055/0.050
$66.00USD / Each
Asco 411M3312HV Solenoid Valve, Three-Way Universal Operation, 12V, 2 Watts, 30 psi EW-98515-38 Mfr # 411M3312HV
COMPARE
0.070/0.033 #10-32 UNF 0.080/0.050
$66.00USD / Each
Asco 411M1324HV Solenoid Valve, Three-Way Universal Operation, 24V, 0.65 Watts, 100 psi EW-98515-40 Mfr # 411M1324HV
COMPARE
0.013/0.008 #10-32 UNF 0.025/0.025
$66.00USD / Each
Asco 411M2324HV Solenoid Valve, Three-Way Universal Operation, 24V, 2 Watts, 50 psi EW-98515-42 Mfr # 411M2324HV
COMPARE
0.038/0.03 #10-32 UNF 0.055/0.050
$66.00USD / Each
Asco 411M3324HV Solenoid Valve, Three-Way Universal Operation, 24V, 2 Watts, 30 psi EW-98515-44 Mfr # 411M3324HV
COMPARE
0.070/0.033 #10-32 UNF 0.080/0.050
$66.00USD / Each
Showing 1- 12 of 12
Need Help with this product?
Call Us At1-847-549-7600
Live chat