Ashcroft® Model 1009 Stainless Steel Pressure Gauges
Ideal for boilers, equipment skids, or severe ambient conditions
  • Meets ASME B40.100 specifications
  • Corrosion resistant stainless steel case
  • Dry gauges are field fillable
  • PowerFlex™ movement provides superior resistance to shock, vibration and pulsation
  • True Zero™ reduces reading errors by using “zero box” instead of conventional dial pins
  • MSL helium leak tested to 1X10-6 scc/s
  • Ventable fill plug
The Ashcroft® type 1009 Industrial Pressure Gauge is ideal for resisting chemical, weather or corrosion effects. Providing durability, reliability and performance, it is well-suited for aggressive process media applications. A good choice for satisfying many application and installation requirements. Available in 2.5" and 3.5" dial sizes, Ashcroft Duralife pressure gauges are liquid fillable and field convertible for panel mounting. Both zero and span adjustments are standard.
ITEM(S) ON SALE
$72.80 - $106.40USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ashcroft 1009 Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 30" Hg to 0 psi EW-68022-10 Mfr # 251009SW 02L 0/30 INHG VAC
COMPARE
Stainless Steel Pressure Gauge, 2.5\" Dial, 1/4\" NPT (M) Lower Connection, 30\" Hg to 0 psi
$74.40USD / Each
$93.00USD / Each
Ashcroft 1009 Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 0 to 30 psi EW-68022-11 Mfr # 251009SW 02L 0/30#
COMPARE
Stainless Steel Pressure Gauge, 2.5\" Dial, 1/4\" NPT (M) Lower Connection, 0 to 30 psi
$74.40USD / Each
$93.00USD / Each
Ashcroft 1009 Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 0 to 200 psi EW-68022-13 Mfr # 251009SW 02L0/200#
COMPARE
Stainless Steel Pressure Gauge, 2.5\" Dial, 1/4\" NPT (M) Lower Connection, 0 to 200 psi
$72.80USD / Each
$91.00USD / Each
Ashcroft 1009 Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 0 to 1000 psi EW-68022-14 Mfr # 251009SW 02L 0/1000#
COMPARE
Stainless Steel Pressure Gauge, 2.5\" Dial, 1/4\" NPT (M) Lower Connection, 0 to 1000 psi
$72.80USD / Each
$91.00USD / Each
Ashcroft 1009 Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 0 to 15 psi EW-68425-63 Mfr # 25-1009-SW-02L-15#
COMPARE
Stainless Steel Pressure Gauge, 2.5\" Dial, 1/4\" NPT (M) Lower Connection, 0 to 15 psi
$74.40USD / Each
$93.00USD / Each
Ashcroft 1009 Stainless Steel Pressure Gauge, 2.5" Dial, 1/4" NPT (M) Lower Connection, 30" Hg to 100 psi EW-68425-64 Mfr # 25-1009-SW-02L-30IHGV/100#
COMPARE
Stainless Steel Pressure Gauge, 2.5\" Dial, 1/4\" NPT (M) Lower Connection, 30\" Hg to 100 psi
$74.40USD / Each
$93.00USD / Each
Ashcroft 1009 Stainless Steel Pressure Gauge, 3.5" Dial, 1/4" NPT (M) Lower Connection, 0 to 5000 psi EW-68425-74 Mfr # 35-1009-SW-02L-5000#
COMPARE
Stainless Steel Pressure Gauge, 3.5\" Dial, 1/4\" NPT (M) Lower Connection, 0 to 5000 psi
$106.40USD / Each
$133.00USD / Each
Showing 1- 7 of 7