Compact 2” gauge size is ideal for limited space applications
Self-draining case is designed for wash down applications
Minimize process dead-leg with small diaphragm
For use with 3/4" tri-clamp connections
Constructed of durable 316L stainless steel wetted parts
Autoclavable
The Ashcroft® Type 1032 sanitary pressure gauge is specifically designed for applications in the food, pharmaceutical and biotechnical industries where small size and sanitary conditions are a priority. This compact autoclavable 2" gauge features all stainless steel construction, temperature-vented case, built-in pressure dampening and a self-draining case to facilitate wash downs. The gauges can also be cleaned or steamed in place.