Atago Digital Pocket Salt Meter
Compact durable unit provides fast and accurate salinity readings
  • Process more samples with the quick 3-second stabilization
  • Measure up to 10% salt concentration in liquid solutions
  • Program the offset feature with dilution or conversion factors for greater accuracy
  • IP65-rated housing offers easy cleanup
This lightweight and compact salt meter is best suited for the food production and service industries. Whether on the plant floor, in a quality control laboratory, or in a busy kitchen, measurements are easy to take and quick to appear on the display.
ITEM(S) ON SALE
$397.89 - $758.25USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Salinity Range (%) Resolution Description
Availability Pricing
Atago PAL-SALT PROBE Salinity Meter, 0 to 7.0% salt concentration EW-02941-31 Mfr # 4222
COMPARE
7 0.01% (0.00 to 1.99%); 0.1% (2.0 to 7.0%) Salinity Meter, 0 to 7.0% salt concentration
$758.25USD / Each
$842.50USD / Each
Atago PAL-SALT Cup Style Salinity Meter, 0 to 10% salt concentration
Always in Stock
EW-02941-32 Mfr # 4250
COMPARE
10 0.01% (0.00 to 2.99%); 0.1% (3.0 to 10.0%) Cup Style Salinity Meter, 0 to 10% salt concentration
$397.89USD / Each
$442.10USD / Each