ATAGO Refractometer
Measure serum protein, specific gravity of urine, and refractive index – with one instrument!
Use these refractometers to provide precise and accurate readings for urine specific gravity, serum protein, and refractive index. Units feature a rugged, high impact plastic design that is corrosion resistant. The large easy-to-read display is screen shaded and the rubber eye guard allows you to comfortably take your measurements. Read your measurements in seconds after placing only a few drops of sample on the prism.
$298.50 - $451.60USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Refractive Index Interval Resolution
Availability Pricing
Atago MASTER-URC/NM Refractometer, 1.000 to 1.050 urine SG, 1.333 to 1.356 nD refractive index EW-02942-05 Mfr # 2793 MASTER-URC/NM
COMPARE
±0.001 (s.g. urine); ±0.001 (nD) 0.001 (s.g. urine); 0.001 (nD)
$298.50USD / Each
Atago MASTER-SUR/NM Refractometer; 1.000-1.060 Urine SG/0.0-12.0 Serum Protein EW-02942-09 Mfr # 2773 MASTER-SUR/NM
COMPARE
±0.001 (s.g. urine); ±0.2 (g serum protein); ±0.0005 (RI) 0.001 (s.g. urine); 0.2 (g serum protein); 0.0005 (RI)
$328.70USD / Each
Atago MASTER-SUR/Na Refractometer; 1.000-1.060 Urine SG/0.0-12.0 Serum Protein, ATC EW-02942-11 Mfr # 2771 MASTER-SUR/Na
COMPARE
±0.001 (s.g. urine); ±0.2 (g serum protein); ±0.0005 (RI) 0.001 (s.g. urine); 0.2 (g serum protein); 0.0005 (RI)
$451.60USD / Each